Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAATTCTCTTTCATGTTAGCTACC[A/G]ATTGCAGAACCAAACGATGTTCTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606342 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SIX4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SIX4 - SIX homeobox 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017420.4 | 2210 | Missense Mutation | TCG,TTG | S,L 717 | NP_059116.3 | |
XM_005267759.2 | 2210 | Missense Mutation | TCG,TTG | S,L 709 | XP_005267816.1 |