Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCTTAGGTCTTAGTACCCACTGT[G/T]TCTATGGTTGGAAAGCATTTGGGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612427 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RBM25 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RBM25 - RNA binding motif protein 25 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021239.2 | 382 | Silent Mutation | GTG,GTT | V,V 58 | NP_067062.1 | |
XM_011537044.2 | 382 | Silent Mutation | GTG,GTT | V,V 58 | XP_011535346.1 | |
XM_011537045.2 | 382 | UTR 5 | XP_011535347.1 |