Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGACGGCAAGCAGCAGAGGAATA[G/T]CACATCGGCCCAACTCACCAGATCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
NOXRED1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NOXRED1 - NADP dependent oxidoreductase domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001113475.2 | 498 | Missense Mutation | ATA,CTA | I,L 140 | NP_001106946.1 | |
XM_005267330.3 | 498 | Missense Mutation | ATA,CTA | I,L 140 | XP_005267387.1 | |
XM_011536427.2 | 498 | Missense Mutation | ATA,CTA | I,L 140 | XP_011534729.1 | |
XM_011536428.2 | 498 | Missense Mutation | ATA,CTA | I,L 140 | XP_011534730.1 | |
XM_011536429.2 | 498 | Missense Mutation | ATA,CTA | I,L 50 | XP_011534731.1 | |
XM_017020969.1 | 498 | Missense Mutation | ATA,CTA | I,L 140 | XP_016876458.1 | |
XM_017020970.1 | 498 | Intron | XP_016876459.1 |