Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCACTTCTACGGTGTCAGTGCCTTT[A/G]CCATTGACTGGCTGTCCATGTGCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610865 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FLVCR2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FLVCR2 - feline leukemia virus subgroup C cellular receptor family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195283.1 | 740 | Intron | NP_001182212.1 | |||
NM_017791.2 | 740 | Missense Mutation | ACC,GCC | T,A 122 | NP_060261.2 |
LOC102724153 - uncharacterized LOC102724153 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |