Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACTGCTGCTCGCGATGGGTGGGGA[C/T]GCCTGGGTGCTCTGACTCTGGGAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613515 | ||||||||||||||||||||
Literature Links: |
ATG14 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ATG14 - autophagy related 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014924.4 | 1123 | Silent Mutation | GCA,GCG | A,A 461 | NP_055739.2 | |
XM_011536563.2 | 1123 | Silent Mutation | GCA,GCG | A,A 348 | XP_011534865.1 |