Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGCTTGGAGAGCTTGATGCTGAAC[A/T]CCTTCCGGCAGTTGTGGCAGTGGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616372 MIM: 605471 | ||||||||||||||||||||
Literature Links: |
DCAF4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DCAF4 - DDB1 and CUL4 associated factor 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZFYVE1 - zinc finger FYVE-type containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281734.1 | 1037 | Missense Mutation | GAG,GTG | E,V 713 | NP_001268663.1 | |
NM_001281735.1 | 1037 | Missense Mutation | GAG,GTG | E,V 312 | NP_001268664.1 | |
NM_021260.3 | 1037 | Missense Mutation | GAG,GTG | E,V 727 | NP_067083.1 | |
NM_178441.2 | 1037 | Missense Mutation | GAG,GTG | E,V 312 | NP_848535.1 | |
XM_017021373.1 | 1037 | Missense Mutation | GAG,GTG | E,V 312 | XP_016876862.1 |