Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTAGGGTAATACCTGCCTGGAGAA[A/T]GGATCTTTCTTACTGAACTTTACAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608577 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHURC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CHURC1 - churchill domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204063.1 | 195 | Missense Mutation | AAA,AAT | K,N 47 | NP_001190992.1 | |
NM_001204064.1 | 195 | Missense Mutation | AAA,AAT | K,N 47 | NP_001190993.1 | |
NM_145165.3 | 195 | Missense Mutation | AAA,AAT | K,N 47 | NP_660148.3 |
CHURC1-FNTB - CHURC1-FNTB readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001202558.1 | 195 | UTR 5 | NP_001189487.1 | |||
NM_001202559.1 | 195 | Missense Mutation | AAA,AAT | K,N 47 | NP_001189488.1 |