Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCTGCTCTGACCTTAGGTAGAGTC[A/C]TCCACAGAAGCATCAAAGTGGACTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 180440 | ||||||||||||||||||||
Literature Links: |
RNASE1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RNASE1 - ribonuclease A family member 1, pancreatic | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002933.4 | 636 | Missense Mutation | GAG,GAT | E,D 153 | NP_002924.1 | |
NM_198232.2 | 636 | Missense Mutation | GAG,GAT | E,D 153 | NP_937875.1 | |
NM_198234.2 | 636 | Missense Mutation | GAG,GAT | E,D 153 | NP_937877.1 | |
NM_198235.2 | 636 | Missense Mutation | GAG,GAT | E,D 153 | NP_937878.1 |