Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAATTACAGTTCAGTTTTAGGGAG[A/T]CAAAGGAAATGAACTTCGGGTATAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615435 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ERO1A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ERO1A - endoplasmic reticulum oxidoreductase 1 alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014584.2 | 3022 | UTR 3 | NP_055399.1 | |||
XM_011536684.2 | 3022 | UTR 3 | XP_011534986.1 |
GPR137C - G protein-coupled receptor 137C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |