Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTTCTATGGATATTATTGATTCC[C/G]TGAGGAAGTAAGTTACATTTCATAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606028 MIM: 603837 | ||||||||||||||||||||
Literature Links: |
CPSF2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CPSF2 - cleavage and polyadenylation specific factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001322270.1 | 410 | Missense Mutation | CTG,GTG | L,V 48 | NP_001309199.1 | |
NM_001322271.1 | 410 | UTR 5 | NP_001309200.1 | |||
NM_001322272.1 | 410 | Missense Mutation | CTG,GTG | L,V 48 | NP_001309201.1 | |
NM_017437.2 | 410 | Missense Mutation | CTG,GTG | L,V 48 | NP_059133.1 |
NDUFB1 - NADH:ubiquinone oxidoreductase subunit B1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |