Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGAAATGTTAGCCAACTCATGGAA[C/T]TGCCTTCCAAAACACTTACTTCACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611280 MIM: 608378 | ||||||||||||||||||||
Literature Links: |
KLHDC2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KLHDC2 - kelch domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014315.2 | 1285 | Silent Mutation | AAC,AAT | N,N 384 | NP_055130.1 | |
XM_006720094.3 | 1285 | Intron | XP_006720157.1 | |||
XM_011536610.1 | 1285 | Missense Mutation | AAC,AAT | N,N 312 | XP_011534912.1 |
NEMF - nuclear export mediator factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_011537317.2 | 1285 | Intron | XP_011535619.1 | |||
XM_011537318.2 | 1285 | Intron | XP_011535620.1 | |||
XM_017021760.1 | 1285 | UTR 3 | XP_016877249.1 | |||
XM_017021761.1 | 1285 | UTR 3 | XP_016877250.1 |