Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTAGGCTTTCTGTGAGGATGAGCA[A/G]CACCGGGATAGACAGGAAGGGCGTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615194 | ||||||||||||||||||||
Literature Links: |
DHRS2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DHRS2 - dehydrogenase/reductase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318835.1 | 196 | Missense Mutation | AAC,AGC | N,S 25 | NP_001305764.1 | |
NM_005794.3 | 196 | Missense Mutation | AAC,AGC | N,S 25 | NP_005785.1 | |
NM_182908.4 | 196 | Missense Mutation | AAC,AGC | N,S 25 | NP_878912.1 | |
XM_005267249.1 | 196 | Missense Mutation | AAC,AGC | N,S 25 | XP_005267306.1 | |
XM_006720001.3 | 196 | Missense Mutation | AAC,AGC | N,S 25 | XP_006720064.1 | |
XM_011536338.1 | 196 | Intron | XP_011534640.1 | |||
XM_011536339.2 | 196 | Intron | XP_011534641.1 |