Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
AGACACTTCTGGTGTGGCCAGACCA[C/T]CTGGGCAGGATTTTGCATCTGGTAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 191050 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
WARS PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
WARS - tryptophanyl-tRNA synthetase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WDR25 - WD repeat domain 25 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161476.1 | 236 | Missense Mutation | CCT,TCT | P,S 46 | NP_001154948.1 | |
NM_024515.4 | 236 | Missense Mutation | CCT,TCT | P,S 46 | NP_078791.3 | |
XM_005268056.4 | 236 | Missense Mutation | CCT,TCT | P,S 46 | XP_005268113.1 | |
XM_005268057.3 | 236 | Intron | XP_005268114.1 | |||
XM_005268058.4 | 236 | Intron | XP_005268115.1 | |||
XM_011537150.2 | 236 | Missense Mutation | CCT,TCT | P,S 46 | XP_011535452.1 | |
XM_011537151.2 | 236 | Intron | XP_011535453.2 | |||
XM_017021639.1 | 236 | Intron | XP_016877128.1 |