Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCATGATCCCTTCTGTGTCTGCG[C/T]GTCCGAATCCCCGTGTGACCCTGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 123875 MIM: 601183 MIM: 603526 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C14orf80 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C14orf80 - chromosome 14 open reading frame 80 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CRIP1 - cysteine rich protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CRIP2 - cysteine rich protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270837.1 | 1390 | UTR 3 | NP_001257766.1 | |||
NM_001270841.1 | 1390 | UTR 3 | NP_001257770.1 | |||
NM_001312.3 | 1390 | UTR 3 | NP_001303.1 |
MTA1 - metastasis associated 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |