Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGATGCCGGACTTGAGCTTCTCTC[C/T]GGTTTGCCCCCCTCTTCCTTGCCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602219 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SALL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SALL2 - spalt like transcription factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291446.1 | 2974 | Silent Mutation | CCA,CCG | P,P 734 | NP_001278375.1 | |
NM_001291447.1 | 2974 | Silent Mutation | CCA,CCG | P,P 732 | NP_001278376.1 | |
NM_005407.2 | 2974 | Silent Mutation | CCA,CCG | P,P 869 | NP_005398.2 | |
XM_011537064.1 | 2974 | Silent Mutation | CCA,CCG | P,P 869 | XP_011535366.1 | |
XM_011537065.1 | 2974 | Silent Mutation | CCA,CCG | P,P 867 | XP_011535367.1 |