Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGAACTCTCACTGGAACTGCTGC[A/G]CGTCTCGGTACTGGTGCTAGTCTTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603607 | ||||||||||||||||||||
Literature Links: |
RPS6KA5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RPS6KA5 - ribosomal protein S6 kinase A5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001322227.1 | 2373 | Missense Mutation | CGC,TGC | R,C 475 | NP_001309156.1 | |
NM_001322228.1 | 2373 | Missense Mutation | CGC,TGC | R,C 720 | NP_001309157.1 | |
NM_001322229.1 | 2373 | Missense Mutation | CGC,TGC | R,C 749 | NP_001309158.1 | |
NM_001322230.1 | 2373 | Missense Mutation | CGC,TGC | R,C 489 | NP_001309159.1 | |
NM_001322231.1 | 2373 | Missense Mutation | CGC,TGC | R,C 517 | NP_001309160.1 | |
NM_001322232.1 | 2373 | Missense Mutation | CGC,TGC | R,C 683 | NP_001309161.1 | |
NM_001322233.1 | 2373 | Missense Mutation | CGC,TGC | R,C 702 | NP_001309162.1 | |
NM_001322234.1 | 2373 | Missense Mutation | CGC,TGC | R,C 564 | NP_001309163.1 | |
NM_001322235.1 | 2373 | Missense Mutation | CGC,TGC | R,C 677 | NP_001309164.1 | |
NM_001322236.1 | 2373 | Missense Mutation | CGC,TGC | R,C 728 | NP_001309165.1 | |
NM_001322237.1 | 2373 | Missense Mutation | CGC,TGC | R,C 677 | NP_001309166.1 | |
NM_001322238.1 | 2373 | Missense Mutation | CGC,TGC | R,C 537 | NP_001309167.1 | |
NM_004755.3 | 2373 | Missense Mutation | CGC,TGC | R,C 756 | NP_004746.2 | |
NM_182398.2 | 2373 | Intron | NP_872198.1 | |||
XM_017021786.1 | 2373 | Missense Mutation | CGC,TGC | R,C 537 | XP_016877275.1 |