Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTAAAGAAATAATTAAGTGAAAAGA[C/T]ATTCAGGTATCCTTTGTTCTCAATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615902 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM177A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM177A1 - family with sequence similarity 177 member A1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC101927178 - uncharacterized LOC101927178 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PPP2R3C - protein phosphatase 2 regulatory subunit B''gamma | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001305155.1 | 1551 | Missense Mutation | ATC,GTC | I,V 253 | NP_001292084.1 | |
NM_001305156.1 | 1551 | Missense Mutation | ATC,GTC | I,V 253 | NP_001292085.1 | |
NM_017917.3 | 1551 | Missense Mutation | ATC,GTC | I,V 363 | NP_060387.2 | |
XM_005267782.3 | 1551 | Missense Mutation | ATC,GTC | I,V 363 | XP_005267839.1 | |
XM_017021388.1 | 1551 | Intron | XP_016876877.1 | |||
XM_017021389.1 | 1551 | Missense Mutation | ATC,GTC | I,V 311 | XP_016876878.1 |