Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAAGCAGCAGAGGAATATCACATC[A/G]GCCCAACTCACCAGATCAGCGTTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
NOXRED1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NOXRED1 - NADP dependent oxidoreductase domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001113475.2 | 491 | Silent Mutation | NP_001106946.1 | |||
XM_005267330.3 | 491 | Silent Mutation | XP_005267387.1 | |||
XM_011536427.2 | 491 | Silent Mutation | XP_011534729.1 | |||
XM_011536428.2 | 491 | Silent Mutation | XP_011534730.1 | |||
XM_011536429.2 | 491 | Silent Mutation | XP_011534731.1 | |||
XM_017020969.1 | 491 | Silent Mutation | XP_016876458.1 | |||
XM_017020970.1 | 491 | Intron | XP_016876459.1 |