Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGGCTCGATGGGGCTCCTGGTGGT[C/T]ACCGAGGCGCTGCCTAGAGGCAGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602294 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FOXA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FOXA1 - forkhead box A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004496.3 | 2042 | Silent Mutation | GTA,GTG | V,V 444 | NP_004487.2 | |
XM_017021246.1 | 2042 | Silent Mutation | GTA,GTG | V,V 411 | XP_016876735.1 |
TTC6 - tetratricopeptide repeat domain 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |