Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATCTACTTTTATTCTTATGTTTTC[A/C]TTCCCAGGAGTGGACCTACCCTATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615814 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
STYX PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
STYX - serine/threonine/tyrosine interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130701.1 | Intron | NP_001124173.1 | ||||
NM_145251.3 | Intron | NP_660294.1 | ||||
XM_006720242.3 | Intron | XP_006720305.1 | ||||
XM_011537108.2 | Intron | XP_011535410.1 | ||||
XM_011537109.2 | Intron | XP_011535411.1 |