Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGAATAGGCAACCAAACCACAATC[A/G]GCTTGTACATTAGCCAAATATTGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CEP128 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CEP128 - centrosomal protein 128 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152446.3 | Intron | NP_689659.2 | ||||
XM_006720056.3 | Intron | XP_006720119.1 | ||||
XM_011536490.2 | Intron | XP_011534792.1 | ||||
XM_011536491.2 | Intron | XP_011534793.1 | ||||
XM_011536492.2 | Intron | XP_011534794.1 | ||||
XM_011536493.2 | Intron | XP_011534795.1 | ||||
XM_011536495.2 | Intron | XP_011534797.1 | ||||
XM_017021042.1 | Intron | XP_016876531.1 | ||||
XM_017021043.1 | Intron | XP_016876532.1 |