Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTATCCAAAGGAAACTTGGAGTTGT[C/T]ACCTAGGGCAGAAAAAAAGGAAAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608249 | ||||||||||||||||||||
Literature Links: |
CCNB1IP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCNB1IP1 - cyclin B1 interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021178.4 | 1123 | Missense Mutation | AAC,GAC | N,D 212 | NP_067001.3 | |
NM_182849.2 | 1123 | Missense Mutation | AAC,GAC | N,D 212 | NP_878269.1 | |
NM_182852.3 | 1123 | Missense Mutation | AAC,GAC | N,D 212 | NP_878272.1 |
TTC5 - tetratricopeptide repeat domain 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |