Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGCCTTCCTCCCCACTTTACTGAA[C/T]CTTCAGTGAGTCTTTCTACCTCAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603055 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C14orf178 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C14orf178 - chromosome 14 open reading frame 178 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001173978.1 | 518 | Missense Mutation | CCT,TCT | P,S 77 | NP_001167449.1 | |
NM_174943.3 | 518 | Missense Mutation | CCT,TCT | P,S 107 | NP_777603.1 |
SNW1 - SNW domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |