Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAAGAAAATTATCACCATTATTAT[A/C]ATCAGAGGTAAGAGTGACTTCGCAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
LOC254028 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC254028 - uncharacterized LOC254028 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RNASE11 - ribonuclease A family member 11 (inactive) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145250.3 | 103 | Intron | NP_660293.1 |
RNASE12 - ribonuclease A family member 12 (inactive) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001024822.2 | 103 | Missense Mutation | ATG,ATT | M,I 1 | NP_001019993.1 |