Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCAGACTCAGCCGCACAGACAGC[A/G]CTTCCTGCTGGGGGAAGTCAAAAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605304 MIM: 607439 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR1260A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR1260A - microRNA 1260a | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NGB - neuroglobin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
POMT2 - protein O-mannosyltransferase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013382.5 | 4197 | UTR 3 | NP_037514.2 | |||
XM_011536675.2 | 4197 | UTR 3 | XP_011534977.1 | |||
XM_011536676.2 | 4197 | UTR 3 | XP_011534978.1 | |||
XM_011536677.2 | 4197 | UTR 3 | XP_011534979.1 | |||
XM_011536679.2 | 4197 | UTR 3 | XP_011534981.1 | |||
XM_017021244.1 | 4197 | UTR 3 | XP_016876733.1 | |||
XM_017021245.1 | 4197 | UTR 3 | XP_016876734.1 |