Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATTGCAGATTTTCTGAGTGCACAC[A/G]AGACCATATCTTTCAGTGGCTGCAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
OR4K1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
OR4K1 - olfactory receptor family 4 subfamily K member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004063.2 | 268 | Intron | NP_001004063.2 | |||
XM_011537153.2 | 268 | Intron | XP_011535455.1 |
OR4K5 - olfactory receptor family 4 subfamily K member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005483.1 | 268 | Missense Mutation | AAG,GAG | K,E 90 | NP_001005483.1 |