Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCACCAACAGGAATGTATCCCTC[C/T]GTGCCTCCCACCGGACCACCTCCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAPK1IP1L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAPK1IP1L - mitogen-activated protein kinase 1 interacting protein 1 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144578.3 | 1230 | Silent Mutation | TCC,TCT | S,S 74 | NP_653179.1 | |
XM_011537362.2 | 1230 | Silent Mutation | TCC,TCT | S,S 87 | XP_011535664.2 | |
XM_017021789.1 | 1230 | Silent Mutation | TCC,TCT | S,S 74 | XP_016877278.1 |