Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTTGCAGATTGAGCTACAGAGAA[C/T]ACCTTGAGGTGTCGCAAGTGTCCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603263 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CA12 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CA12 - carbonic anhydrase 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001218.4 | 1513 | UTR 3 | NP_001209.1 | |||
NM_001293642.1 | 1513 | UTR 3 | NP_001280571.1 | |||
NM_206925.2 | 1513 | UTR 3 | NP_996808.1 |