Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAAGCAGTGCTTCCACGAGGAGGT[A/G]GAGCAGGGCGTGAAGTTCTCCCTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMED3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TMED3 - transmembrane p24 trafficking protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301203.1 | 323 | Silent Mutation | GTA,GTG | V,V 45 | NP_001288132.1 | |
NM_007364.3 | 323 | Silent Mutation | GTA,GTG | V,V 45 | NP_031390.1 | |
XM_017022040.1 | 323 | Silent Mutation | GTA,GTG | V,V 45 | XP_016877529.1 | |
XM_017022041.1 | 323 | Silent Mutation | GTA,GTG | V,V 45 | XP_016877530.1 |