Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGGTGCATCCCAGCGTAGGCCTGC[C/T]GCAGGGGGTCAGCCACGCCGGGGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612681 | ||||||||||||||||||||
Literature Links: |
CELF6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CELF6 - CUGBP, Elav-like family member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172684.1 | 1133 | Missense Mutation | CAG,CGG | Q,R 354 | NP_001166155.1 | |
NM_001172685.1 | 1133 | Intron | NP_001166156.1 | |||
NM_052840.4 | 1133 | Missense Mutation | CAG,CGG | Q,R 354 | NP_443072.3 |