Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTCACTGGAAGATGAAAAAGCCA[A/G]TTTTGCCCACAAAAGAGTTTGGGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607342 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CPEB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CPEB1 - cytoplasmic polyadenylation element binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001079533.1 | Intron | NP_001073001.1 | ||||
NM_001079534.1 | Intron | NP_001073002.1 | ||||
NM_001079535.1 | Intron | NP_001073003.1 | ||||
NM_001288819.1 | Intron | NP_001275748.1 | ||||
NM_001288820.1 | Intron | NP_001275749.1 | ||||
NM_030594.4 | Intron | NP_085097.3 |