Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGTTCTAAAAGGATGGTTCAGTT[C/T]CCTCACCCTGCCTAACCCAAGGCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147521 MIM: 151520 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ITPKA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ITPKA - inositol-trisphosphate 3-kinase A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002220.2 | Intron | NP_002211.1 | ||||
XM_011521522.2 | Intron | XP_011519824.1 |
LTK - leukocyte receptor tyrosine kinase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |