Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGAAGATCCACCCACCGGGGGAA[C/T]GCAGGGCTCCCCAGTACACGTTTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNM1P35 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DNM1P35 - dynamin 1 pseudogene 35 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ODF3L1 - outer dense fiber of sperm tails 3 like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_175881.3 | 628 | Missense Mutation | CGC,TGC | R,C 136 | NP_787077.1 | |
XM_006720414.3 | 628 | Missense Mutation | CGC,TGC | R,C 153 | XP_006720477.1 |