Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAATGCCATACCTGATCCTGGTGA[C/T]CAAAGAAGGTGTGAACACAAGTCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609540 | ||||||||||||||||||||
Literature Links: |
DNAJA4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DNAJA4 - DnaJ heat shock protein family (Hsp40) member A4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WDR61 - WD repeat domain 61 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303247.1 | 1019 | Missense Mutation | GAT,GGT | D,G 272 | NP_001290176.1 | |
NM_001303248.1 | 1019 | Missense Mutation | GAT,GGT | D,G 179 | NP_001290177.1 | |
NM_025234.2 | 1019 | Missense Mutation | GAT,GGT | D,G 272 | NP_079510.1 | |
XM_011522094.1 | 1019 | Missense Mutation | GAT,GGT | D,G 272 | XP_011520396.1 | |
XM_017022637.1 | 1019 | UTR 3 | XP_016878126.1 |