Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAACAGAGATTGGCTCAGGATGGA[A/G]TTTGTTCAGTTTCAGTTCTGCAGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147370 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IGF1R PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IGF1R - insulin like growth factor 1 receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000875.4 | Intron | NP_000866.1 | ||||
NM_001291858.1 | Intron | NP_001278787.1 | ||||
XM_011521516.2 | Intron | XP_011519818.1 | ||||
XM_011521517.2 | Intron | XP_011519819.1 | ||||
XM_017022136.1 | Intron | XP_016877625.1 | ||||
XM_017022137.1 | Intron | XP_016877626.1 | ||||
XM_017022138.1 | Intron | XP_016877627.1 | ||||
XM_017022139.1 | Intron | XP_016877628.1 |
IRAIN - IGF1R antisense imprinted non-protein coding RNA | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |