Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCAGCCTTCAGGGAAGAGGAGGA[A/C]TGGGCCAGCAGAACTGGCTGGAAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616567 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DAPK2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DAPK2 - death associated protein kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014326.3 | Intron | NP_055141.2 | ||||
XM_011521413.2 | Intron | XP_011519715.1 | ||||
XM_011521414.1 | Intron | XP_011519716.1 | ||||
XM_011521415.2 | Intron | XP_011519717.1 | ||||
XM_011521416.2 | Intron | XP_011519718.1 | ||||
XM_011521417.1 | Intron | XP_011519719.1 | ||||
XM_011521421.2 | Intron | XP_011519723.1 | ||||
XM_017022043.1 | Intron | XP_016877532.1 | ||||
XM_017022044.1 | Intron | XP_016877533.1 | ||||
XM_017022045.1 | Intron | XP_016877534.1 | ||||
XM_017022046.1 | Intron | XP_016877535.1 | ||||
XM_017022047.1 | Intron | XP_016877536.1 | ||||
XM_017022048.1 | Intron | XP_016877537.1 | ||||
XM_017022049.1 | Intron | XP_016877538.1 | ||||
XM_017022050.1 | Intron | XP_016877539.1 | ||||
XM_017022051.1 | Intron | XP_016877540.1 | ||||
XM_017022052.1 | Intron | XP_016877541.1 |