Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGTTTTATGACTCTCCGTTAACT[C/T]TGCTGTCTCCTACATTAATGCAAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613261 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRTG PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PRTG - protogenin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173814.5 | 10507 | UTR 3 | NP_776175.2 | |||
XM_011521459.1 | 10507 | Intron | XP_011519761.1 | |||
XM_011521460.1 | 10507 | Intron | XP_011519762.1 | |||
XM_017022081.1 | 10507 | UTR 3 | XP_016877570.1 | |||
XM_017022082.1 | 10507 | Intron | XP_016877571.1 |