Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCGAATTCTCTCCACCATCTTCTT[T/C]AAAGGCTACAACCATCAAAGTGAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606322 MIM: 608146 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYFIP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CYFIP1 - cytoplasmic FMR1 interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033028.1 | 4146 | Silent Mutation | TTA,TTG | L,L 770 | NP_001028200.1 | |
NM_001287810.2 | 4146 | Silent Mutation | TTA,TTG | L,L 1201 | NP_001274739.1 | |
NM_001324119.1 | 4146 | Silent Mutation | TTA,TTG | L,L 1235 | NP_001311048.1 | |
NM_001324120.1 | 4146 | Silent Mutation | TTA,TTG | L,L 1201 | NP_001311049.1 | |
NM_001324122.1 | 4146 | Silent Mutation | TTA,TTG | L,L 641 | NP_001311051.1 | |
NM_001324123.1 | 4146 | Silent Mutation | TTA,TTG | L,L 1201 | NP_001311052.1 | |
NM_001324124.1 | 4146 | Silent Mutation | TTA,TTG | L,L 1171 | NP_001311053.1 | |
NM_001324125.1 | 4146 | Silent Mutation | TTA,TTG | L,L 1079 | NP_001311054.1 | |
NM_001324126.1 | 4146 | Silent Mutation | TTA,TTG | L,L 1167 | NP_001311055.1 | |
NM_014608.4 | 4146 | Silent Mutation | TTA,TTG | L,L 1201 | NP_055423.1 | |
XM_011543873.2 | 4146 | Silent Mutation | TTA,TTG | L,L 1334 | XP_011542175.1 | |
XM_011543874.1 | 4146 | Silent Mutation | TTA,TTG | L,L 1334 | XP_011542176.1 | |
XM_011543876.2 | 4146 | Silent Mutation | TTA,TTG | L,L 1233 | XP_011542178.2 | |
XM_017022023.1 | 4146 | Silent Mutation | TTA,TTG | L,L 1368 | XP_016877512.1 | |
XM_017022024.1 | 4146 | Silent Mutation | TTA,TTG | L,L 1334 | XP_016877513.1 |
NIPA2 - non imprinted in Prader-Willi/Angelman syndrome 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |