Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCACTCACACCACATCAACTCCTA[C/T]GGACAAAAGGGCTTTTTCTAAGCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610014 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TM2D3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TM2D3 - TM2 domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001307960.1 | 827 | Intron | NP_001294889.1 | |||
NM_001308026.1 | 827 | Intron | NP_001294955.1 | |||
NM_025141.3 | 827 | UTR 3 | NP_079417.2 | |||
NM_078474.2 | 827 | UTR 3 | NP_510883.2 |