Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGAGGGGCTGTGAGGCTCACCCTC[C/T]GCCGCGCTCAGGAGGACCCGCCGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147650 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IDH2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IDH2 - isocitrate dehydrogenase (NADP(+)) 2, mitochondrial | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001289910.1 | 1334 | UTR 3 | NP_001276839.1 | |||
NM_001290114.1 | 1334 | UTR 3 | NP_001277043.1 | |||
NM_002168.3 | 1334 | UTR 3 | NP_002159.2 |
ZNF710 - zinc finger protein 710 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |