Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCATGATATCTCAAATCATTTATA[C/T]ATTTTATCTTTTTTTTTTTTTTTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608706 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DYX1C1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DYX1C1 - dyslexia susceptibility 1 candidate 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033559.2 | Intron | NP_001028731.1 | ||||
NM_001033560.1 | Intron | NP_001028732.1 | ||||
NM_130810.3 | Intron | NP_570722.2 |
DYX1C1-CCPG1 - DYX1C1-CCPG1 readthrough (NMD candidate) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |