Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAGTTCAACTAATGACTTCCCAAC[A/G]TGGGAGGCAAAAAACTTCATGAATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 114240 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CAPN3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CAPN3 - calpain 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF106 - zinc finger protein 106 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001284306.1 | 5458 | UTR 3 | NP_001271235.1 | |||
NM_001284307.1 | 5458 | UTR 3 | NP_001271236.1 | |||
NM_022473.2 | 5458 | UTR 3 | NP_071918.1 | |||
XM_005254591.2 | 5458 | UTR 3 | XP_005254648.1 | |||
XM_005254592.2 | 5458 | UTR 3 | XP_005254649.1 | |||
XM_017022483.1 | 5458 | UTR 3 | XP_016877972.1 | |||
XM_017022484.1 | 5458 | UTR 3 | XP_016877973.1 | |||
XM_017022485.1 | 5458 | UTR 3 | XP_016877974.1 |