Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGAGGACATGTTGAGGGCCAGCAG[A/G]ACAAAGCATATATCCTGGCCCATGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 177070 MIM: 603805 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EPB42 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EPB42 - erythrocyte membrane protein band 4.2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TGM5 - transglutaminase 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004245.3 | 792 | Silent Mutation | GTC,GTT | V,V 444 | NP_004236.1 | |
NM_201631.3 | 792 | Silent Mutation | GTC,GTT | V,V 526 | NP_963925.2 | |
XM_011522230.2 | 792 | Silent Mutation | GTC,GTT | V,V 183 | XP_011520532.1 | |
XM_017022729.1 | 792 | Silent Mutation | GTC,GTT | V,V 183 | XP_016878218.1 |