Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACACAGCATTCAGTTTTGATGCAC[A/G]CTTTTGTAGAAAGCACATAATGGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615795 MIM: 188060 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FSIP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FSIP1 - fibrous sheath interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001324338.1 | Intron | NP_001311267.1 | ||||
NM_152597.4 | Intron | NP_689810.3 | ||||
XM_011521305.2 | Intron | XP_011519607.1 | ||||
XM_011521306.1 | Intron | XP_011519608.1 | ||||
XM_011521307.2 | Intron | XP_011519609.1 | ||||
XM_011521309.2 | Intron | XP_011519611.1 | ||||
XM_011521311.2 | Intron | XP_011519613.1 | ||||
XM_017021972.1 | Intron | XP_016877461.1 |
THBS1 - thrombospondin 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |