Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACTGCCCATGGTCTTCAAGGCCAG[A/G]ATGACCTAGCCCCATTTACCTTTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613766 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SCAMP5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SCAMP5 - secretory carrier membrane protein 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001178111.1 | Intron | NP_001171582.1 | ||||
NM_001178112.1 | Intron | NP_001171583.1 | ||||
NM_138967.3 | Intron | NP_620417.1 | ||||
XM_006720420.1 | Intron | XP_006720483.1 | ||||
XM_006720421.1 | Intron | XP_006720484.1 | ||||
XM_006720422.3 | Intron | XP_006720485.1 | ||||
XM_017021989.1 | Intron | XP_016877478.1 |