Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAAAAAAATACACCTCTCATTAA[C/T]GAAATTTTGCACATAGACATCTGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603158 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
USP8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
USP8 - ubiquitin specific peptidase 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128610.2 | Intron | NP_001122082.1 | ||||
NM_001283049.1 | Intron | NP_001269978.1 | ||||
NM_005154.4 | Intron | NP_005145.3 | ||||
XM_006720761.3 | Intron | XP_006720824.1 | ||||
XM_006720762.3 | Intron | XP_006720825.1 | ||||
XM_011522193.2 | Intron | XP_011520495.1 | ||||
XM_017022718.1 | Intron | XP_016878207.1 | ||||
XM_017022719.1 | Intron | XP_016878208.1 | ||||
XM_017022720.1 | Intron | XP_016878209.1 | ||||
XM_017022721.1 | Intron | XP_016878210.1 | ||||
XM_017022722.1 | Intron | XP_016878211.1 |