Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTGCAACATCTGAAAAGTAGTGC[A/C]TGTAGAAGGAACTTGAAATTGATCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603566 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PIAS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PIAS1 - protein inhibitor of activated STAT 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320687.1 | 138 | Intron | NP_001307616.1 | |||
NM_016166.2 | 138 | Intron | NP_057250.1 | |||
XM_011522126.2 | 138 | Intron | XP_011520428.1 | |||
XM_011522127.2 | 138 | Intron | XP_011520429.1 | |||
XM_017022688.1 | 138 | Intron | XP_016878177.1 | |||
XM_017022689.1 | 138 | Intron | XP_016878178.1 | |||
XM_017022690.1 | 138 | UTR 5 | XP_016878179.1 | |||
XM_017022691.1 | 138 | Intron | XP_016878180.1 |