Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCTCAAAGATGGGCCCCCTTCCT[A/C]CAGGCCCCAAGGGCCCCTTTAACTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605265 MIM: 180903 | ||||||||||||||||||||
Literature Links: |
AVEN PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AVEN - apoptosis and caspase activation inhibitor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020371.2 | 946 | Missense Mutation | NP_065104.1 | |||
XM_011521818.2 | 946 | Missense Mutation | XP_011520120.1 | |||
XM_011521819.2 | 946 | Missense Mutation | XP_011520121.1 | |||
XM_011521820.1 | 946 | Missense Mutation | XP_011520122.1 |
RYR3 - ryanodine receptor 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |