Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCATCCATTGCTCCAGGCCCAGGG[A/G]CCCAGATGGTCTGATTCCCTGTGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612457 MIM: 602990 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARID3B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARID3B - AT-rich interaction domain 3B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CLK3 - CDC like kinase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130028.1 | Intron | NP_001123500.1 | ||||
NM_003992.4 | Intron | NP_003983.2 | ||||
XM_005254151.3 | Intron | XP_005254208.1 | ||||
XM_011521205.2 | Intron | XP_011519507.1 | ||||
XM_011521206.2 | Intron | XP_011519508.1 | ||||
XM_011521209.2 | Intron | XP_011519511.2 | ||||
XM_017021905.1 | Intron | XP_016877394.1 | ||||
XM_017021906.1 | Intron | XP_016877395.1 | ||||
XM_017021907.1 | Intron | XP_016877396.1 | ||||
XM_017021908.1 | Intron | XP_016877397.1 | ||||
XM_017021909.1 | Intron | XP_016877398.1 | ||||
XM_017021910.1 | Intron | XP_016877399.1 |
LOC102723750 - uncharacterized LOC102723750 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_017022789.1 | Intron | XP_016878278.1 | ||||
XM_017022790.1 | Intron | XP_016878279.1 | ||||
XM_017022791.1 | Intron | XP_016878280.1 | ||||
XM_017022792.1 | Intron | XP_016878281.1 | ||||
XM_017022793.1 | Intron | XP_016878282.1 | ||||
XM_017022794.1 | Intron | XP_016878283.1 | ||||
XM_017022795.1 | Intron | XP_016878284.1 | ||||
XM_017022796.1 | Intron | XP_016878285.1 | ||||
XM_017022797.1 | Intron | XP_016878286.1 | ||||
XM_017022798.1 | Intron | XP_016878287.1 |