Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTGCCAGATTTTAAGATTAAACTG[C/T]TTGCCATCTCTTTTTTTTTTCTTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610183 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZFAND6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZFAND6 - zinc finger AN1-type containing 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242911.1 | Intron | NP_001229840.1 | ||||
NM_001242912.1 | Intron | NP_001229841.1 | ||||
NM_001242913.1 | Intron | NP_001229842.1 | ||||
NM_001242914.1 | Intron | NP_001229843.1 | ||||
NM_001242915.1 | Intron | NP_001229844.1 | ||||
NM_001242916.1 | Intron | NP_001229845.1 | ||||
NM_001242917.1 | Intron | NP_001229846.1 | ||||
NM_001242918.1 | Intron | NP_001229847.1 | ||||
NM_001242919.1 | Intron | NP_001229848.1 | ||||
NM_019006.3 | Intron | NP_061879.2 | ||||
XM_011521684.2 | Intron | XP_011519986.1 |